(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 99%
Supplier: Apollo Scientific
Warning
Synonyms: (2,2-Dimethyl-1,3-dioxolan-4-yl)methanol, 2,2-Dimethyl-1,3-dioxolan-4-ylmethanol, DL-1,2-Isopropylidenglycerol, 1,2-O-Isopropylideneglycerol, Solketal, (±)-1,2-O-Isopropylidenglycerol, DL-1,2-Isopropylideneglycerol
OR019104-100G OR019104-500G OR019104-25G
APOSOR019104-100GEA 24.8 EUR
APOSOR019104-100G APOSOR019104-500G APOSOR019104-25G
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 99%
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
(contains isomer)
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 188…189 °C
Density: 1.064
Flash Pt: 90 °C
MDL Number: MFCD00063238
CAS Number: 100-79-8
Merck Index: 13,05233