4'-Chloropropiophenone
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Chlorophenyl)propan-1-one
OR11048-25G OR11048-100G OR11048-500G
APOSOR11048-25GEA 37.7 EUR
APOSOR11048-25G APOSOR11048-100G APOSOR11048-500G
4'-Chloropropiophenone
4'-Chloropropiophenone
Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 125…126/13 mm °C
Melting Pt: 35…37 °C
Flash Pt: >113 °C
MDL Number: MFCD00000626
CAS Number: 6285-05-8