2-Fluorothiophenol 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Fluorobenzenethiol, 1-Fluoro-2-mercaptobenzene, 2-Fluorobenzene-1-thiol, o-Fluorothiophenol
PC4298-25G PC4298-5G PC4298-100G
APOSPC4298-25GEA 72.5 EUR
APOSPC4298-25G APOSPC4298-5G APOSPC4298-100G
2-Fluorothiophenol 98%
2-Fluorothiophenol
Formula: C₆H₅FS
Boiling Pt: 61 - 62/35 mm °C
Density: 1.203
Flash Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041419
CAS Number: 2557-78-0
UN: 3336
ADR: 3,III