2-Bromothiophenol 97%
Supplier: Apollo Scientific
Danger
OR2222-5G OR2222-25G
APOSOR2222-5GEA 21.9 EUR
APOSOR2222-5G APOSOR2222-25G
2-Bromothiophenol 97%
2-Bromothiophenol
Formula: C₆H₅BrS
Boiling Pt: 128 - 130/25 mm °C
Density: 1.573
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004827
CAS Number: 6320-02-1
UN: 2922
ADR: 8,III