4-Fluoro-3-(trifluoromethyl)benzaldehyde 90%
Supplier: Apollo Scientific
Warning
PC99378-25G PC99378-100G
APOSPC99378-25GEA 40.6 EUR
APOSPC99378-25G APOSPC99378-100G
4-Fluoro-3-(trifluoromethyl)benzaldehyde 90%
4-Fluoro-3-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₄O
MW: 192.11 g/mol
Boiling Pt: 60 °C
Density: 1.408
Flash Pt: 80 °C
MDL Number: MFCD00061314
CAS Number: 67515-60-0