L(-)-Proline
Supplier: Thermo Scientific
SB02034.1 SB02034.10
MAYBSB02034.1EA 0 EUR
MAYBSB02034.1 MAYBSB02034.10
L(-)-Proline
L(-)-Proline
Formula: C₅H₉NO₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 220…222 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064318
CAS Number: 147-85-3
EINECS: 205-702-2
Merck Index: 13,07871