trans-4-Hydroxy-L(-)-proline, Sigma-Aldrich®
Supplier: Merck
Synonyms:
L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
56250-100G
56250-25G
56250-5G
SIAL56250-100GEA
449
EUR
SIAL56250-100G
SIAL56250-25G
SIAL56250-5G
trans-4-Hydroxy-L(-)-proline, Sigma-Aldrich®
trans-4-Hydroxy-L(-)-proline
Formula:
C₅H₉NO₃ MW: 131.13 g/mol Boiling Pt: 296 Melting Pt: 206 Storage Temperature: Ambient |
MDL Number:
MFCD00064320 CAS Number: 51-35-4 EINECS: 200-091-9 Merck Index: 13,04866 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...