trans-4-Hydroxy-L(-)-proline, Sigma-Aldrich®
Supplier: Merck
Synonyms: L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
M-Clarity™ quality level = MQ200
H5534-100G H5534-10MG H5534-1KG H5534-25G
SIALH5534-100GEA 1140 EUR
SIALH5534-100G SIALH5534-10MG SIALH5534-1KG SIALH5534-25G
trans-4-Hydroxy-L(-)-proline, Sigma-Aldrich®
trans-4-Hydroxy-L(-)-proline
Formula: C₅H₉NO₃
MW: 131.13 g/mol
Boiling Pt: 296
Melting Pt: 206
Storage Temperature: Ambient
MDL Number: MFCD00064320
CAS Number: 51-35-4
EINECS: 200-091-9
Merck Index: 13,04866