JALAS® Tempus 5668 ESD Safety Shoes, BOA® Fit System
Supplier: EJENDALS
5668-41 5668-42 5668-43 5668-44 5668-45 5668-46 5668-47 5668-48 5668-35 5668-36 5668-37 5668-38 5668-39 5668-40
EJEN5668-41EA 180 EUR
EJEN5668-41 EJEN5668-42 EJEN5668-43 EJEN5668-44 EJEN5668-45 EJEN5668-46 EJEN5668-47 EJEN5668-48 EJEN5668-35 EJEN5668-36 EJEN5668-37 EJEN5668-38 EJEN5668-39 EJEN5668-40
JALAS® Tempus 5668 ESD Safety Shoes, BOA® Fit System
Shoes and Boots
Safety low shoes for both work and leisure. Very light and comfortable with green details, safe and sporty with BOA® Fit system.
  • Anti-static
  • Protective steel toecap
  • Oil-resistant outsole
  • Ergothan advanced shock absorption system with PORON XRD
  • Nail protection in plasma-treated composite (PTC) textile material
The well proven outsole has a very effective grip pattern and is made of a special nitrile rubber for high durability. The JALAS® Tempus shoes are ESD and antistatic certified to be used in ESD protected work environments.
Outer sole made of nitrile rubber, uppper material of microfiber textile and FX2 pro slim insole.
Certifications: EN ISO 20345:2011, S1, P, SRC; ASTM F2413-18 M/I/C SD 35 PR, ASTM F3445-21 SR; DGUV 112-191.