Furfuryl alcohol 98%
Supplier: Thermo Fisher Scientific
Danger
119790010 119790025 119790100 119792500
ACRO119790010EA 61.9 EUR
ACRO119790010 ACRO119790025 ACRO119790100 ACRO119792500
Furfuryl alcohol 98%
Furfuryl alcohol
Formula: C₅H₆O₂
Boiling Pt: 170 °C (760 mmHg)
Melting Pt: –29 °C
Density: 1.13 g/cm³
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00003252
CAS Number: 98-00-0
EINECS: 202-626-1
UN: 2874
ADR: 6.1,III
Merck Index: 13,04326
Order Now
Specification Test Results
Appearance Clear yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4850 to 1.4880 (20°C, 589 nm)
Water ≤0.3 %
GC ≥97.5 %