Pentaerythritol 98%
Supplier: Thermo Fisher Scientific
129870010 129872500 129870050
ACRO129870010EA 37.7 EUR
ACRO129870010 ACRO129872500 ACRO129870050
Pentaerythritol 98%
Pentaerythritol
Formula: C₅H₁₂O₄
Boiling Pt: 276 °C (30 mmHg)
Melting Pt: 257…264 °C
Density: 1,511 g/cm³ (20 °C)
Flash Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00004692
CAS Number: 115-77-5
EINECS: 204-104-9
Merck Index: 12,07245
Order Now
Specification Test Results
Appearance White Crystals
Infrared spectrum Conforms
Melting point 257 to 264 °C
GC ≥97.5 %