Tetraphenylcyclopentadienone 99%
Supplier: Thermo Fisher Scientific
138540250
138541000
138540050
ACRO138540250EA
106
EUR
ACRO138540250
ACRO138541000
ACRO138540050
Tetraphenylcyclopentadienone 99%
Tetraphenylcyclopentadienone
Formula:
C₂₉H₂₀O Melting Pt: 217…222 °C Density: 1,233 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001407 CAS Number: 479-33-4 EINECS: 207-530-3 |
Specification Test Results
Appearance | Black Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 217 to 222 °C |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...