2-Methylbenzothiazole 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
150381000
150385000
150380250
ACRO150381000EA
156
EUR
ACRO150381000
ACRO150385000
ACRO150380250
2-Methylbenzothiazole 99%
2-Methylbenzothiazole
Formula:
C₈H₇NS Boiling Pt: 238 °C (760 mmHg) Melting Pt: 12…14 °C Density: 1.2 g/cm³ Flash Pt: 102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005794 CAS Number: 120-75-2 EINECS: 204-423-3 |
Specification Test Results
Appearance | Clear colorless to yellow to orange-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6150 to 1.6180 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...