2-Naphthalenesulphonyl chloride 97%

Supplier: Thermo Scientific
Danger

AC15844.1 AC15844.10 AC15844.50
MAYBAC15844.1EA 0 EUR
MAYBAC15844.1 MAYBAC15844.10 MAYBAC15844.50
2-Naphthalenesulphonyl chloride 97%
2-Naphthalenesulphonyl chloride
Formula: C₁₀H₇ClO₂S
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 74…78 °C
MDL Number: MFCD00004087
CAS Number: 93-11-8
EINECS: 202-219-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 74 to 78 °C
Titration Argentometric ≥96.0 % (ex Cl)
Residue after ignition ≤0.005 %
Chloride (Cl) ≤1 ppm
Titrable acid ≤0.0005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR