2-Amino-5-nitrothiazole 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
5-Nitrothiazol-2-ylamine, 5-Nitrothiazol-2-amine
158521000
158520250
ACRO158521000EA
84.1
EUR
ACRO158521000
ACRO158520250
2-Amino-5-nitrothiazole 97%
2-Amino-5-nitrothiazole
Formula:
C₃H₃N₃O₂S Melting Pt: 196…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005326 CAS Number: 121-66-4 EINECS: 204-490-9 Merck Index: 13,00456 |
Specification Test Results
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Identification | Passes test |
Relative density | 0.790 to 0.793 |
Residue after evaporation | ≤50 ppm |
Related substances | ≤2 ppm (Benzene) |
Acidity/alkalinity | Passes test |
Insoluble matter | (in water) Passes test |
Reducing substances | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...