1-Chloroethyl chloroformate 97+%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 1-Chloroethyl carbonochloridate
314460250 314461000
ACRO314460250EA 79.6 EUR
ACRO314460250 ACRO314461000
1-Chloroethyl chloroformate 97+%
1-Chloroethyl chloroformate
Formula: C₃H₄Cl₂O₂
MW: 142.97 g/mol
Boiling Pt: 118…119 °C
Melting Pt: -65
Density: 1.32 g/cm³
Flash Pt: 40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000647
CAS Number: 50893-53-3
EINECS: 256-834-2
UN: 2742
ADR: 6.1 II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4240 (20°C, 589 nm)
GC ≥97.0 %