Allyltrimethylsilane 97%
Supplier: Thermo Fisher Scientific
|
Danger
|
349990100
349990500
ACRO349990100EA
44.7
EUR
ACRO349990100
ACRO349990500
Allyltrimethylsilane 97%
Allyltrimethylsilane
Formula:
H₂CCHCH₂Si(CH₃)₃ MW: 114.26 g/mol Boiling Pt: 84 Density: 0.719 g/cm³ Flash Pt: 7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008635 CAS Number: 762-72-1 EINECS: 212-104-5 UN: 1993 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4060 to 1.4080 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...