Sodium nitroprusside dihydrate ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Sodium pentacyanonitrosylferrate(III) dihydrate, Sodium nitrosopentacyanoferrate (III) dihydrate, Sodium nitroferricyanide(III) dihydrate, Disodium pentacyanonitrosylferrate (III) dihydrate, Sodium nitroferricyanide dihydrate
424360250 424361000 424365000 424360025
ACRO424360250EA 45.7 EUR
ACRO424360250 ACRO424361000 ACRO424365000 ACRO424360025
Sodium nitroprusside dihydrate ACS, meets analytical specifications of Reagent Ph. Eur.
Sodium nitroprusside dihydrate
Formula: Na₂[Fe(CN)₅NO]·2H₂O
MW: 297.95 g/mol
Density: 1.72 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149192
CAS Number: 13755-38-9
EINECS: 238-373-9
UN: 1588
ADR: 6.1 III
Merck Index: 13,08722
Order Now
Specification Test Results
Appearance Bordeaux Crystals
Titration Argentometric 99.0 to 102.0 %
Chloride (Cl) ≤0.02 %
Sulfate (SO4) Passes test (Limit about 0.01%)
Insoluble matter ≤0.01 %