Diammonium tetrachloroplatinate (≥51% Pt) ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Ammonium tetrachloroplatinate(II), Ammonium tetrachloroplatinate
11046.03 11046.06
11046.03EA 147 EUR
11046.03 11046.06
Diammonium tetrachloroplatinate (≥51% Pt) ≥99.9% (metals basis)
Diammonium tetrachloroplatinate
Purity: 99.9% CAS No.: 13820-41-2 Molecular Formula: H8Cl4N2Pt Formula Weight: 372.98 Physical Form: Crystalline Density: 2.936 Appearance: Dark red Odour: Odourless Melting Point: 145° dec. UN No.: 3288 MDL No.: MFCD00010885
Formula: H₈Cl₄N₂Pt
MW: 372.97 g/mol
Melting Pt: 145 °C (dec.)
Density: 2.936
Storage Temperature: Ambient
MDL Number: MFCD00010885
CAS Number: 13820-41-2
EINECS: 237-499-1
UN: 3288
ADR: 6.1 III
Merck Index: 12,00583