trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 97.5-100.5% ACS
Supplier: Thermo Fisher Scientific
|
Warning
|
36697.06
36697.14
36697.22
36697.36
36697.06EA
39.9
EUR
36697.06
36697.14
36697.22
36697.36
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 97.5-100.5% ACS
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
Purity: 97.5%-100.5%
CAS No.: 125572-95-4
Molecular Formula: C14H22N2O8.H2O
Formula Weight: 364.35 (346.34anhy)
Physical Form: Solid
Melting Point: 213-216°
Beilstein Reference: 3222748
MDL No.: MFCD00066429
Formula:
C₁₄H₂₂N₂O₈·1H₂O Melting Pt: 213…216 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066429 CAS Number: 125572-95-4 EINECS: 236-308-9 |