Tetraphenylcyclopentadienone ≥98%
Supplier: Thermo Fisher Scientific
A12756.06
A12756.14
A12756.22
A12756.06EA
35.9
EUR
A12756.06
A12756.14
A12756.22
Tetraphenylcyclopentadienone ≥98%
Tetraphenylcyclopentadienone
Purity: 98%
CAS No.: 479-33-4
Molecular Formula: C29H20O
Formula Weight: 384.48
Melting Point: 217-219°
Beilstein Reference: 683115
Formula:
C₂₉H₂₀O Melting Pt: 217…219 °C Density: 1.233 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001407 CAS Number: 479-33-4 EINECS: 207-530-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...