Triphenylsilanol ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A19284.06
A19284.14
A19284.22
AB125413.0025
A19284.06EA
23.5
EUR
A19284.06
A19284.14
A19284.22
ABCRAB125413.0025
Triphenylsilanol ≥98%
Triphenylsilanol
Purity: 98%
CAS No.: 791-31-1
Molecular Formula: C18H16OSi
Formula Weight: 276.41
Melting Point: 149-154°
Beilstein Reference: 985007
Formula:
C₁₈H₁₆OSi Melting Pt: 149…154 °C Density: 1.17 Storage Temperature: Ambient |
MDL Number:
MFCD00002102 CAS Number: 791-31-1 EINECS: 212-339-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...