ω-Chloro-2,4-difluoroacetophenone ≥98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 2-Chloro-1-(2,4-difluorophenyl)ethanone, α-Chloro-2,4-difluoroacetophenone, 2-Chloro-2',4'-difluoroacetophenone
B22487.06 B22487.14
B22487.06EA 61.6 EUR
B22487.06 B22487.14
ω-Chloro-2,4-difluoroacetophenone ≥98%
ω-Chloro-2,4-difluoroacetophenone
Purity: 98% CAS No.: 51336-94-8 Molecular Formula: C8H5ClF2O Formula Weight: 190.58 Flash Point: 110°(230°F) Melting Point: 44-48° UN No.: 3261 MDL No.: MFCD00013252
Formula: C₈H₅ClF₂O
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: 44…48 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013252
CAS Number: 51336-94-8
UN: 3261
ADR: 8,II