Tetrabromophthalic anhydride ≥98%
Supplier: Thermo Fisher Scientific
Warning
B24550.22 B24550.36
B24550.22EA 46.4 EUR
B24550.22 B24550.36
Tetrabromophthalic anhydride ≥98%
Tetrabromophthalic anhydride
Purity: 98% CAS No.: 632-79-1 Molecular Formula: C8Br4O3 Formula Weight: 463.70 Odour: Odourless Melting Point: 274-280° Beilstein Reference: 18908 MDL No.: MFCD00005919
Formula: C₈Br₄O₃
Boiling Pt: 477 °C (1013 hPa)
Melting Pt: 274…280 °C
Density: 2.87
Storage Temperature: Ambient
MDL Number: MFCD00005919
CAS Number: 632-79-1
EINECS: 211-185-4