2-Bromo-3-(trifluoromethyl)pyridine ≥98%
Supplier: Thermo Fisher Scientific
Danger
H25822.06 H25822.03
H25822.06EA 338 EUR
H25822.06 H25822.03
2-Bromo-3-(trifluoromethyl)pyridine ≥98%
2-Bromo-3-(trifluoromethyl)pyridine
Purity: 98% CAS No.: 175205-82-0 Molecular Formula: C6H3BrF3N Formula Weight: 226.00 Flash Point: 29°(84°F) Melting Point: 37-39° Boiling Point: 76°/8mm MDL No.: MFCD00153084
Formula: C₆H₃BrF₃N
MW: 226 g/mol
Boiling Pt: 76 °C (8 mmHg)
Melting Pt: 37…39 °C
Flash Pt: 29 °C (84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00153084
CAS Number: 175205-82-0
UN: 2926
ADR: 4.1 III