1-Chloro-4-ethynylbenzene ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
H51058.03
H51058.06
H51058.03EA
176
EUR
H51058.03
H51058.06
1-Chloro-4-ethynylbenzene ≥98%
1-Chloro-4-ethynylbenzene
Purity: 98%
CAS No.: 873-73-4
Molecular Formula: C8H5Cl
Formula Weight: 136.58
Flash Point: 57°(135°F)
Melting Point: 43-47°
Boiling Point: 179°
UN No.: 1325
MDL No.: MFCD00191917
Formula:
C₈H₅Cl Boiling Pt: 179 °C Melting Pt: 43…47 °C Flash Pt: 57 °C (135 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00191917 CAS Number: 873-73-4 UN: 1325 ADR: 4.1,III |