2-Chloro-4-(trifluoromethyl)pyridine ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
L10713.03
L10713.06
L10713.03EA
88.3
EUR
L10713.03
L10713.06
2-Chloro-4-(trifluoromethyl)pyridine ≥98%
2-Chloro-4-(trifluoromethyl)pyridine
Purity: 98+%
CAS No.: 81565-18-6
Molecular Formula: C6H3ClF3N
Formula Weight: 181.55
Flash Point: 40°(104°F)
Refractive Index: 1.4490
Boiling Point: 145°
Beilstein Reference: 4246276
UN No.: 1992
MDL No.: MFCD00042224
Formula:
C₆H₃ClF₃N MW: 181.54 g/mol Boiling Pt: 145 °C Melting Pt: –1 °C Density: 1.411 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042224 CAS Number: 81565-18-6 UN: 1992 ADR: 3 III |