Sodium hypochlorite (11 - 14% Cl₂) in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
L14709.0D L14709.0F L14709.AP
L14709.0DEA 222 EUR
L14709.0D L14709.0F L14709.AP
Sodium hypochlorite (11 - 14% Cl₂) in aqueous solution
Sodium hypochlorite
Formula: ClNaO
Boiling Pt: 111 °C (1013 hPa)
Melting Pt: –30…–20 °C
Density: 1.22…1.25 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8,II