2,3-Diaminotoluene
Supplier: Thermo Scientific
Synonyms:
3-Methylbenzene-1,2-diamine, Toluene-2,3-diamine, 1,2-Diamino-3-methylbenzene
BTB07447DA
BTB07447EA
BTB07447.50
MAYBBTB07447DAEA
0
EUR
MAYBBTB07447DA
MAYBBTB07447EA
MAYBBTB07447.50
2,3-Diaminotoluene
2,3-Diaminotoluene
Formula:
C₇H₁₀N₂ MW: 122.17 g/mol Boiling Pt: 255 Melting Pt: 113 Density: 1.3 g/cm³ (20 °C) |
MDL Number:
MFCD00011589 CAS Number: 2687-25-4 EINECS: 220-248-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...