ω-Bromo-4-methoxyacetophenone
Supplier: Thermo Scientific
Warning
Synonyms: 4-Methoxyphenacyl bromide, Bromomethyl p-methoxyphenyl ketone, α-Bromo-4-methoxyacetophenone, 2-Bromo-1-(4-methoxyphenyl)ethan-1-one, 2-Bromo-1-(4-methoxyphenyl)ethanone, 2-Bromo-4'-methoxyacetophenone
CD00356.1 CD00356EA CD00356.50
MAYBCD00356.1EA 0 EUR
MAYBCD00356.1 MAYBCD00356EA MAYBCD00356.50
ω-Bromo-4-methoxyacetophenone
ω-Bromo-4-methoxyacetophenone
Formula: C₉H₉BrO₂
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 67…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000201
CAS Number: 2632-13-5
EINECS: 220-118-8
UN: 3261
ADR: 8,II