DL-Camphor
Supplier: Thermo Scientific
Warning
NRB04998.250 NRB04998.1
MAYBNRB04998.250EA 0 EUR
MAYBNRB04998.250 MAYBNRB04998.1
DL-Camphor
DL-Camphor
Formula: C₁₀H₁₆O
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: ∼180 °C
Density: 0.992 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00074738
CAS Number: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III