1-(3-Iodopropoxy)-4-chlorobenzene

Supplier: Thermo Scientific

RH01447DA RH01447EA
MAYBRH01447DAEA 0 EUR
MAYBRH01447DA MAYBRH01447EA
1-(3-Iodopropoxy)-4-chlorobenzene
1-(3-Iodopropoxy)-4-chlorobenzene
Formula: C₉H₁₀ClIO
CAS Number: 119795-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR