Diethylene glycol monobutyl ether, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 2-(2-Butoxyethoxy)ethanol, Diethylene glycol mono-n-butyl ether, Butyldigol

M-Clarity™ quality level = MQ200

579963-1L 579963-18L 579963-4L
SIAL579963-1LEA 43.8 EUR
SIAL579963-1L SIAL579963-18L SIAL579963-4L
Diethylene glycol monobutyl ether, Sigma-Aldrich®
Diethylene glycol monobutyl ether
Formula: CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
Boiling Pt: 230.4 °C (1013 hPa)
Melting Pt: –68 °C
Density: 0.954 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002881
CAS Number: 112-34-5
EINECS: 203-961-6
Merck Index: 13,01556

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR