1-Ethyl-3-methylimidazoliumtetrafluoroborate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium tetrafluoroborate
M-Clarity™ quality level = MQ100
04365-50G 04365-5G
SIAL04365-50GEA 334 EUR
SIAL04365-50G SIAL04365-5G
1-Ethyl-3-methylimidazoliumtetrafluoroborate, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium tetrafluoroborate
Formula: C₆H₁₁BF₄N₂
MW: 197.97 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: ∼15 °C
Density: 1.34 g/cm³ (20 °C)
Flash Pt: >110 °C
MDL Number: MFCD00216668
CAS Number: 143314-16-3