Vinyltrichlorosilane, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

104876-100G 104876-500G
SIAL104876-100GEA 37.3 EUR
SIAL104876-100G SIAL104876-500G
Vinyltrichlorosilane, Sigma-Aldrich®
Vinyltrichlorosilane
Formula: C₂H₃Cl₃Si
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.2426 g/cm³ (20 °C)
MDL Number: MFCD00000482
CAS Number: 75-94-5
EINECS: 200-917-8
UN: 1305
ADR: 3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR