Methyl chloroacetate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
108413-1KG 108413-500G
SIAL108413-1KGEA 91.5 EUR
SIAL108413-1KG SIAL108413-500G
Methyl chloroacetate, Sigma-Aldrich®
Methyl chloroacetate
Formula: C₃H₅ClO₂
MW: 108.52 g/mol
Boiling Pt: 131
Melting Pt: –33 °C
Density: 1.2378 g/cm³ (20 °C)
Flash Pt: 47
MDL Number: MFCD00000931
CAS Number: 96-34-4
EINECS: 202-501-1
UN: 2295
ADR: 6.1 I
Merck Index: 12,06122