Thiazole, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
151645-1G 151645-5G
SIAL151645-1GEA 38.9 EUR
SIAL151645-1G SIAL151645-5G
Thiazole, Sigma-Aldrich®
Thiazole
Formula: C₃H₃NS
MW: 85.13 g/mol
Boiling Pt: 116.8
Melting Pt: –34 °C
Density: 1.2 g/cm³ (20 °C)
MDL Number: MFCD00005315
CAS Number: 288-47-1
EINECS: 206-021-3
Merck Index: 12,09442