Isovaleryl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
157422-100G 157422-25G 157422-500G
SIAL157422-100GEA 67 EUR
SIAL157422-100G SIAL157422-25G SIAL157422-500G
Isovaleryl chloride, Sigma-Aldrich®
Isovaleryl chloride
Formula: C₅H₉ClO
Boiling Pt: 115 °C (1013 hPa)
Density: 0.99 g/cm³ (20 °C)
MDL Number: MFCD00000738
CAS Number: 108-12-3
EINECS: 203-552-2
UN: 2920
ADR: 8,II
Merck Index: 13,05253