4-Biphenylcarbonyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
161144-10G 161144-50G
SIAL161144-10GEA 67.1 EUR
SIAL161144-10G SIAL161144-50G
4-Biphenylcarbonyl chloride, Sigma-Aldrich®
4-Biphenylcarbonyl chloride
Formula: C₁₃H₉ClO
Boiling Pt: 195…200 °C (5 mmHg)
Melting Pt: 110…112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000692
CAS Number: 14002-51-8
EINECS: 237-804-8
UN: 3261
ADR: 8,II