Adipoyl dichloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Adipoyl chloride

M-Clarity™ quality level = MQ200

165212-100G 165212-25G 165212-5G
SIAL165212-100GEA 155 EUR
SIAL165212-100G SIAL165212-25G SIAL165212-5G
Adipoyl dichloride, Sigma-Aldrich®
Adipoyl dichloride
Formula: C₆H₈Cl₂O₂
Boiling Pt: 125 °C (15 hPa)
Density: 1.2563 g/cm³ (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00000759
CAS Number: 111-50-2
EINECS: 203-876-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR