Tris(triphenylphosphine)rhodium(I) chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Wilkinson's catalyst, Chlorotris(triphenylphosphine)rhodium(I), Tris(triphenylphosphine)chlororhodium
M-Clarity™ quality level = MQ100
205036-1G 205036-250MG 205036-5G
SIAL205036-1GEA 162 EUR
SIAL205036-1G SIAL205036-250MG SIAL205036-5G
Tris(triphenylphosphine)rhodium(I) chloride, Sigma-Aldrich®
Tris(triphenylphosphine)rhodium (I) chloride
Formula: C₅₄H₄₅ClP₃Rh
MW: 925.23 g/mol
Melting Pt: 157
Density: 1.363 g/cm³ (20 °C)
MDL Number: MFCD00010016
CAS Number: 14694-95-2
EINECS: 238-744-5
Merck Index: 12,02226