Bis(2-butoxyethyl)ether, Sigma-Aldrich®
Supplier: Merck
Synonyms: Diethylene glycol dibutyl ether, 2,2-Dibutoxyethyl ether
M-Clarity™ quality level = MQ200
205621-100ML 205621-1L 205621-500ML
SIAL205621-100MLEA 57 EUR
SIAL205621-100ML SIAL205621-1L SIAL205621-500ML
Bis(2-butoxyethyl)ether, Sigma-Aldrich®
Bis(2-butoxyethyl) ether
Formula: C₁₂H₂₆O₃
MW: 218.34 g/mol
Boiling Pt: 256
Melting Pt: –60 °C
Density: 0.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009459
CAS Number: 112-73-2
EINECS: 204-001-9