Allyltrimethylsilane, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

208264-10G 208264-50G
SIAL208264-10GEA 263 EUR
SIAL208264-10G SIAL208264-50G
Allyltrimethylsilane, Sigma-Aldrich®
Allyltrimethylsilane
Formula: H₂CCHCH₂Si(CH₃)₃
Boiling Pt: 84 °C (1013 hPa)
Density: 0.7139 g/cm³ (20 °C)
MDL Number: MFCD00008635
CAS Number: 762-72-1
EINECS: 212-104-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR