α,α,α-Trifluoro-4-tolualdehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4-(Trifluoromethyl)benzaldehyde, 1-Formyl-4-trifluoromethylbenzene, p-Trifluoromethylbenzaldehyde
224944-10G
224944-50G
SIAL224944-10GEA
94.3
EUR
SIAL224944-10G
SIAL224944-50G
α,α,α-Trifluoro-4-tolualdehyde, Sigma-Aldrich®
α,α,α-Trifluoro-4-tolualdehyde
Formula:
C₈H₅F₃O |
MDL Number:
MFCD00006952 CAS Number: 455-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...