Ethyl chloroformate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Chloroformic acid ethyl ester
M-Clarity™ quality level = MQ100
23131-100ML 23131-1L 23131-500ML
SIAL23131-100MLEA 56.4 EUR
SIAL23131-100ML SIAL23131-1L SIAL23131-500ML
Ethyl chloroformate, Sigma-Aldrich®
Ethyl chloroformate
Formula: C₃H₅ClO₂
Boiling Pt: 93 °C (1013 hPa)
Melting Pt: –81 °C
Density: 1.14 g/cm³ (20 °C)
Flash Pt: 10 °C
MDL Number: MFCD00000644
CAS Number: 541-41-3
EINECS: 208-778-5
UN: 1182
ADR: 6.1,I
Merck Index: 12,03831