N-Phenyl-p-phenylenediamine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Aminodiphenylamine, N-(4-Aminophenyl)aniline

M-Clarity™ quality level = MQ100

241393-100G 241393-1KG
SIAL241393-100GEA 50.8 EUR
SIAL241393-100G SIAL241393-1KG
N-Phenyl-p-phenylenediamine, Sigma-Aldrich®
N-Phenyl-p-phenylenediamine
Formula: C₁₂H₁₂N₂
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 72…75 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 193 °C
MDL Number: MFCD00007850
CAS Number: 101-54-2
EINECS: 202-951-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR