1,2-Diacetylbenzene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
242039-100MG 242039-500MG
SIAL242039-100MGEA 84.5 EUR
SIAL242039-100MG SIAL242039-500MG
1,2-Diacetylbenzene, Sigma-Aldrich®
1,2-Diacetylbenzene
Formula: C₁₀H₁₀O₂
Boiling Pt: 348 °C (1013 hPa)
Melting Pt: 38…39 °C
CAS Number: 704-00-7
EINECS: 211-877-6