Neopentyl bromide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
249890-25G 249890-5G
SIAL249890-25GEA 198 EUR
SIAL249890-25G SIAL249890-5G
Neopentyl bromide, Sigma-Aldrich®
Neopentyl bromide
Formula: C₅H₁₁Br
Boiling Pt: 106 °C (1013 hPa)
Density: 1.2253 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000209
CAS Number: 630-17-1
EINECS: 211-132-5
UN: 1993
ADR: 3,II