N-(Bromomethyl)phthalimide, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-(Bromomethyl)isoindoline-1,3-dione
M-Clarity™ quality level = MQ100
252611-25G 252611-5G
SIAL252611-25GEA 201 EUR
SIAL252611-25G SIAL252611-5G
N-(Bromomethyl)phthalimide, Sigma-Aldrich®
N-(Bromomethyl)phthalimide
Formula: C₉H₆BrNO₂
MW: 240.06 g/mol
Boiling Pt: 410
Melting Pt: 148…151 °C
MDL Number: MFCD00005897
CAS Number: 5332-26-3
EINECS: 226-239-2