Oil of castor, Sigma-Aldrich®
Supplier: Merck
Synonyms: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi
M-Clarity™ quality level = MQ200
259853-250ML 259853-1L
SIAL259853-250MLEA 38.2 EUR
SIAL259853-250ML SIAL259853-1L
Oil of castor, Sigma-Aldrich®
Oil of castor
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: –10 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: >113 °C (closed cup)
MDL Number: MFCD00130746
CAS Number: 8001-79-4
EINECS: 232-293-8
Merck Index: 13,01908